Thiosalicylic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Thiobenzoic acid, 2-Mercaptobenzoic acid
A13401.0B
A13401.18
A13401.30
A13401.0BEA
471
GBP
A13401.0B
A13401.18
A13401.30
Thiosalicylic acid ≥98%
Thiosalicylic acid
Purity: 98%
CAS No.: 147-93-3
Molecular Formula: C7H6O2S
Formula Weight: 154.19
Odour: Stench
Melting Point: 165-168°
Beilstein Reference: 508507
Merck Reference: 14,9360
MDL No.: MFCD00004836
Formula:
C₇H₆O₂S MW: 154.19 g/mol Boiling Pt: 230 °C (0.02 mmHg) Melting Pt: 165…168 °C Density: 1.49 Storage Temperature: Ambient |
MDL Number:
MFCD00004836 CAS Number: 147-93-3 EINECS: 205-704-3 UN: 3335 ADR: 9, Merck Index: 14,9360 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...