2,3-Dimethoxybenzoic acid

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

MOLEM85593190EA 0 GBP
MOLEM85593190
2,3-Dimethoxybenzoic acid
2,3-Dimethoxybenzoic acid
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 121…124 °C
Density: 1,387 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002432
CAS Number: 1521-38-6
EINECS: 216-188-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR