Anthranilonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
023851-250G
023851-50G
FLUO023851-250GEA
0
GBP
FLUO023851-250G
FLUO023851-50G
Anthranilonitrile
Anthranilonitrile
Formula:
C₇H₆N₂ Boiling Pt: 268 °C (1013 hPa) Melting Pt: 47 °C Density: 1.26 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007631 CAS Number: 1885-29-6 EINECS: 217-549-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...