Salicylaldehyde

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Hydroxybenzaldehyde

MOLE30786630EA 0 GBP
MOLE30786630
Salicylaldehyde
Salicylaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –7 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003317
CAS Number: 90-02-8
EINECS: 201-961-0
Merck Index: 13,08405

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR