Vanillin, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Hydroxy-3-methoxybenzaldehyde
M-Clarity™ quality level = MQ200
SIAL94752-100GEA
54.3
GBP
SIAL94752-100G
Vanillin, Sigma-Aldrich®
Vanillin
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 285 °C (1013 hPa) Melting Pt: ∼82 °C Density: 1.056 g/cm³ (20 °C) Flash Pt: 153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006942 CAS Number: 121-33-5 EINECS: 204-465-2 Merck Index: 13,09998 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...