2-Bromo-1,4-dimethoxybenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Bromo-2,5-dimethoxybenzene
003694-25G
003694-100G
FLUO003694-25GEA
0
GBP
FLUO003694-25G
FLUO003694-100G
2-Bromo-1,4-dimethoxybenzene
2-Bromo-1,4-dimethoxybenzene
Formula:
C₈H₉BrO₂ MW: 217.06 g/mol Boiling Pt: 107 °C (0.2 mmHg) Density: 1,445 g/cm³ (15 °C) |
MDL Number:
MFCD00008355 CAS Number: 25245-34-5 EINECS: 246-756-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...