3-Bromoaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003072-100G
003072-500G
FLUO003072-100GEA
0
GBP
FLUO003072-100G
FLUO003072-500G
3-Bromoaniline
3-Bromoaniline
Formula:
C₆H₆BrN MW: 172.02 g/mol Boiling Pt: 251 °C (1013 hPa) Melting Pt: 16 °C Density: 1.59 g/cm³ (20 °C) Flash Pt: >230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007757 CAS Number: 591-19-5 EINECS: 209-704-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...