2,4-Dibromoaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003660-25G
003660-100G
FLUO003660-25GEA
0
GBP
FLUO003660-25G
FLUO003660-100G
2,4-Dibromoaniline
2,4-Dibromoaniline
Formula:
C₆H₅Br₂N MW: 250.92 g/mol Boiling Pt: 156 °C (21 mmHg) Melting Pt: 80…82 °C Density: 2.26 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007633 CAS Number: 615-57-6 EINECS: 210-434-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...