2,6-Dibromo-4-fluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Dibromo-5-fluoro-2-hydroxybenzene

013295-25G 013295-5G
FLUO013295-25GEA 0 GBP
FLUO013295-25G FLUO013295-5G
2,6-Dibromo-4-fluorophenol
2,6-Dibromo-4-fluorophenol
Formula: C₆H₃Br₂FO
MW: 269.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002317
CAS Number: 344-20-7
EINECS: 206-451-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR