1-Bromo-4-(1,1,2,2-tetrafluoroethoxy)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Bromo-alpha,alpha,beta,beta-tetrafluorophenetole

010258-25G 010258-5G
FLUO010258-25GEA 0 GBP
FLUO010258-25G FLUO010258-5G
1-Bromo-4-(1,1,2,2-tetrafluoroethoxy)benzene
1-Bromo-4-(1,1,2,2-tetrafluoroethoxy)benzene
Formula: C₈H₅BrF₄O
MW: 273.02 g/mol
MDL Number: MFCD00042190
CAS Number: 68834-05-9
EINECS: 272-431-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR