1-(Trimethylsilyl)-1-pentyne

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SILS19980.5 SILS19980.25
SILNSILS19980.5EA 0 GBP
SILNSILS19980.5 SILNSILS19980.25
1-(Trimethylsilyl)-1-pentyne
1-(Trimethylsilyl)-1-pentyne
Formula: C₈H₁₆Si
MW: 140.3 g/mol
MDL Number: MFCD00077890
CAS Number: 18270-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR