3-Iodo-4-methoxybenzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Iodo-p-anisic acid

717800-25G 717800-5G
SIAL717800-25GEA 260 GBP
SIAL717800-25G SIAL717800-5G
3-Iodo-4-methoxybenzoic acid, Sigma-Aldrich®
3-Iodo-4-methoxybenzoic acid
Formula: C₈H₇IO₃
MW: 278.05 g/mol
MDL Number: MFCD00017347
CAS Number: 68507-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR