Sodium dihydrogen citrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate
71497-1KG
71497-250G
SIAL71497-1KGEA
376
GBP
SIAL71497-1KG
SIAL71497-250G
Sodium dihydrogen citrate, Sigma-Aldrich®
Sodium dihydrogen citrate
Formula:
C₆H₇NaO₇ MW: 214.11 g/mol Melting Pt: 209…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013067 CAS Number: 18996-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...