3-Bromo-4-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004210-5G 004210-25G 004210-100G
FLUO004210-5GEA 0 GBP
FLUO004210-5G FLUO004210-25G FLUO004210-100G
3-Bromo-4-fluorobenzonitrile
3-Bromo-4-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 134…136 °C (33 mmHg)
Melting Pt: 53…58 °C
MDL Number: MFCD00055432
CAS Number: 79630-23-2
EINECS: 279-200-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR