2-(Biphenyl-4-yl)-2-propanol

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: α,α-Dimethyl[1,1'-biphenyl]-4-methanol2-(4-Biphenylyl)-2-propanol

8.01965.0010EA 0 GBP
8.01965.9999 8.01965.0010
2-(Biphenyl-4-yl)-2-propanol
2-(Biphenyl-4-yl)-2-propanol
Formula: C₁₅H₁₆O
MW: 212.29 g/mol
MDL Number: MFCD00014390
CAS Number: 34352-74-4
EINECS: 251-955-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR