4-(Trifluoromethyl)phenylhydrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(4-(Trifluoromethyl)phenyl)hydrazine, 4-Hydrazinobenzotrifluoride

007647-5G 007647-25G
FLUO007647-5GEA 0 GBP
FLUO007647-5G FLUO007647-25G
4-(Trifluoromethyl)phenylhydrazine
4-(Trifluoromethyl)phenylhydrazine
Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
MDL Number: MFCD00042508
CAS Number: 368-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR