2,3,5,6-Tetrafluorophenylhydrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007370-5G 007370-25G
FLUO007370-5GEA 0 GBP
FLUO007370-5G FLUO007370-25G
2,3,5,6-Tetrafluorophenylhydrazine
2,3,5,6-Tetrafluorophenylhydrazine
Formula: C₆H₄F₄N₂
MDL Number: MFCD00007575
CAS Number: 653-11-2
EINECS: 211-494-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR