1H,1H-Pentafluoropropyl methacrylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1H,1H-Pentafluoropropyl methacrylate, 2,2,3,3,3-Pentafluoropropyl Methacrylate, Methacrylic acid 2,2,2,3,3-pentafluoropropyl ester, Methacrylic acid 2,2,3,3,3-Pentafluoropropyl ester, 1H,1H-Perfluoropropyl methacrylate

006988-5G 006988-25G
FLUO006988-5GEA 0 GBP
FLUO006988-5G FLUO006988-25G
1H,1H-Pentafluoropropyl methacrylate
1H,1H-Pentafluoropropyl methacrylate
Formula: C₇H₇F₅O₂
MW: 218.12 g/mol
MDL Number: MFCD00039256
CAS Number: 45115-53-5
EINECS: 256-191-8
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR