(Ethylsulphonyl)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001288-5G 001288-25G
FLUO001288-5GEA 0 GBP
FLUO001288-5G FLUO001288-25G
(Ethylsulphonyl)benzene
(Ethylsulphonyl)benzene
Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
MDL Number: MFCD00025081
CAS Number: 599-70-2
EINECS: 209-972-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR