2,4-Diamino-6-phenyl-1,3,5-triazine
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Phenyl-1,3,5-triazine-2,4-diyldiamine
, 6-Phenyl-1,3,5-triazine-2,4-diamine
, 2,4-Diamino-6-phenyl-s-triazine
, Benzoguanamine
, 2,6-Diamino-4-phenyl-1,3,5-triazine
EHERC10537450EA
135
GBP
EHERC10537450
2,4-Diamino-6-phenyl-1,3,5-triazine
2,4-Diamino-6-phenyl-1,3,5-triazine
Formula:
C₉H₉N₅ MW: 187.2 g/mol |
MDL Number:
MFCD00023187 CAS Number: 91-76-9 EINECS: 202-095-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...