1-Bromo-2,4,6-trifluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Bromo-1,3,5-trifluorobenzene

001799-25G 001799-100G
FLUO001799-25GEA 0 GBP
FLUO001799-25G FLUO001799-100G
1-Bromo-2,4,6-trifluorobenzene
1-Bromo-2,4,6-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 140.5 °C (1013 hPa)
Melting Pt: 3.5 °C
Density: 1.79 g/cm³ (20 °C)
MDL Number: MFCD00000334
CAS Number: 2367-76-2
EINECS: 219-125-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR