1,4-Dibromotetrafluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
004668-5G
004668-25G
004668-100G
FLUO004668-5GEA
0
GBP
FLUO004668-5G
FLUO004668-25G
FLUO004668-100G
1,4-Dibromotetrafluorobenzene
1,4-Dibromotetrafluorobenzene
Formula:
C₆Br₂F₄ MW: 307.87 g/mol Boiling Pt: 198 °C (1013 hPa) Melting Pt: 78.2…78.5 °C |
MDL Number:
MFCD00000309 CAS Number: 344-03-6 EINECS: 206-448-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...