1,2-Dibromotetrafluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,2-Dibromo-3,4,5,6-tetrafluorobenzene
009415-5G
009415-1G
FLUO009415-5GEA
0
GBP
FLUO009415-5G
FLUO009415-1G
1,2-Dibromotetrafluorobenzene
1,2-Dibromotetrafluorobenzene
Formula:
C₆Br₂F₄ MW: 307.87 g/mol Boiling Pt: 93…96 °C (25 mmHg) Melting Pt: 12 °C Density: 2,23 g/cm³ (20 °C) |
MDL Number:
MFCD00039209 CAS Number: 827-08-7 EINECS: 212-564-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...