2-Bromo-5-chlorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018255-1G
018255-5G
018255-25G
FLUO018255-1GEA
0
GBP
FLUO018255-1G
FLUO018255-5G
FLUO018255-25G
2-Bromo-5-chlorobenzoic acid
2-Bromo-5-chlorobenzoic acid
Formula:
C₇H₄BrClO₂ MW: 235.46 g/mol |
MDL Number:
MFCD00013982 CAS Number: 21739-93-5 EINECS: 244-559-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...