1-Bromo-3,5-dichlorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,5-Dichlorobromobenzene

023277-100G 023277-25G
FLUO023277-100GEA 0 GBP
FLUO023277-100G FLUO023277-25G
1-Bromo-3,5-dichlorobenzene
1-Bromo-3,5-dichlorobenzene
Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 82…84 °C
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR