3,5-Dibromo-4-hydroxybenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018500-10G 018500-25G
FLUO018500-10GEA 0 GBP
FLUO018500-10G FLUO018500-25G
3,5-Dibromo-4-hydroxybenzoic acid
3,5-Dibromo-4-hydroxybenzoic acid
Formula: C₇H₄Br₂O₃
MW: 295.91 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 275…276 °C
MDL Number: MFCD00002548
CAS Number: 3337-62-0
EINECS: 222-075-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR