1-Tetradecene

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

EHERC17397100EA 94.4 GBP
EHERC17397100
1-Tetradecene
1-Tetradecene
Formula: C₁₄H₂₈
MW: 196.38 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: –13 °C
Density: 0,7708 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008981
CAS Number: 1120-36-1
EINECS: 214-306-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR