2,3-Dichloropropene

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,3-Dichloro-1-propene

EHERCA12490500EA 94.4 GBP
EHERCA12490500
2,3-Dichloropropene
2,3-Dichloropropene
Formula: C₃H₄Cl₂
MW: 110.97 g/mol
Boiling Pt: 92.5 °C (1013 hPa)
Melting Pt: –83.1 °C
Density: 1.209 g/cm³ (20 °C)
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000943
CAS Number: 78-88-6
EINECS: 201-153-8
UN: 2047
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR