6-Bromo-1-hexene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
013446-10G
013446-50G
FLUO013446-10GEA
0
GBP
FLUO013446-10G
FLUO013446-50G
6-Bromo-1-hexene
6-Bromo-1-hexene
Formula:
C₆H₁₁Br MW: 163.06 g/mol Boiling Pt: 34…36 °C (0.05 mmHg) Density: 1.2169 g/cm³ (20 °C) |
MDL Number:
MFCD00000269 CAS Number: 2695-47-8 EINECS: 220-267-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...