n-Tetracosane

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-tetracosane

EHERC17395800EA 96.8 GBP
EHERC17395800
n-Tetracosane
n-Tetracosane
Formula: H₃C(CH₂)₂₂CH₃
MW: 338.66 g/mol
Boiling Pt: 391.3 °C (1013 hPa)
Melting Pt: 49…51 °C
Density: 0.7787 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009352
CAS Number: 646-31-1
EINECS: 211-474-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR