n-Pentatriacontane, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

76968-250MGEA 64.2 GBP
76968-250MG
n-Pentatriacontane, Supelco®
n-Pentatriacontane
Formula: H₃C(CH₂)₃₃CH₃
MW: 492.96 g/mol
Boiling Pt: 490 °C (1013 hPa)
Melting Pt: 73.8 °C
Density: 0.8157 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00059397
CAS Number: 630-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR