n-Pentadecane
Supplier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N-Pentadecane
, Pentadecane
PROOCHE168EA
287
GBP
PROOCHE168
n-Pentadecane
n-Pentadecane
Formula:
H₃C(CH₂)₁₃CH₃ MW: 212.42 g/mol Boiling Pt: 271 °C (1013 hPa) Melting Pt: 13…15 °C Density: 0.768 g/cm³ (20 °C) Flash Pt: 132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008990 CAS Number: 629-62-9 EINECS: 211-098-1 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...