3-(Trimethylsilyl)propiolic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Trimethylsilyl)propynoic acid

SILS20140.10 SILS20140.25
SILNSILS20140.10EA 0 GBP
SILNSILS20140.10 SILNSILS20140.25
3-(Trimethylsilyl)propiolic acid
3-(Trimethylsilyl)propiolic acid
Formula: C₆H₁₀O₂Si
MW: 142.23 g/mol
MDL Number: MFCD00190216
CAS Number: 5683-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR