Trimethylsilyl propiolate
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Propynoic acid trimethylsilyl ester, Propiolic acid trimethylsilyl ester
SILS20130.1
SILS20130.5
SILNSILS20130.1EA
0
GBP
SILNSILS20130.1
SILNSILS20130.5
Trimethylsilyl propiolate
Trimethylsilyl propiolate
Formula:
C₆H₁₀O₂Si MW: 142.23 g/mol |
CAS Number:
19232-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...