Ethyl 3-(trimethylsilyl)propiolate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 3-(trimethylsilyl)propynoate, 3-(Trimethylsilyl)-2-propynoic acid ethyl ester, 3-(Trimethylsilyl)propiolic acid ethyl ester, Ethyl 3-(trimethylsilyl)propiolate

SILS08905.5 SILS08905.25
SILNSILS08905.5EA 0 GBP
SILNSILS08905.5 SILNSILS08905.25
Ethyl 3-(trimethylsilyl)propiolate
Ethyl 3-(trimethylsilyl)propiolate
Formula: C₈H₁₄O₂Si
MW: 170.28 g/mol
MDL Number: MFCD00074886
CAS Number: 16205-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR