Potassium (4-iodophenyl)trifluoroborate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(4-Iodophenyl)trifluoroboric acid potassium salt, Potassium 4-iodophenyltrifluoroborate
M-Clarity™ quality level = MQ100
706108-1G
706108-5G
SIAL706108-1GEA
127
GBP
SIAL706108-1G
SIAL706108-5G
Potassium (4-iodophenyl)trifluoroborate, Sigma-Aldrich®
Potassium (4-iodophenyl)trifluoroborate
Formula:
C₆H₄BF₃IK MW: 309.91 g/mol |
MDL Number:
MFCD09800738 CAS Number: 912350-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...