2-Bromo-3,5-bis(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Bis(trifluoromethyl)-2-bromoaniline

013770-1G 013770-5G
FLUO013770-1GEA 0 GBP
FLUO013770-1G FLUO013770-5G
2-Bromo-3,5-bis(trifluoromethyl)aniline
2-Bromo-3,5-bis(trifluoromethyl)aniline
Formula: C₈H₄BrF₆N
MW: 308.02 g/mol
MDL Number: MFCD00134419
CAS Number: 174824-16-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR