3,4-Dibromo-1,1,1,2,2-pentafluorobutane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010285-1G 010285-5G
FLUO010285-1GEA 0 GBP
FLUO010285-1G FLUO010285-5G
3,4-Dibromo-1,1,1,2,2-pentafluorobutane
3,4-Dibromo-1,1,1,2,2-pentafluorobutane
Formula: C₄H₃Br₂F₅
MDL Number: MFCD03094110
CAS Number: 378-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR