1H,1H,2H,2H-Heptadecafluoro-1-decanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanol, 1H,1H,2H,2H-Perfluoro-1-decanol, 1H,1H,2H,2H-Perfluorodecan-1-ol
007020-25G
007020-100G
007020-250G
FLUO007020-25GEA
0
GBP
FLUO007020-25G
FLUO007020-100G
FLUO007020-250G
1H,1H,2H,2H-Heptadecafluoro-1-decanol
1H,1H,2H,2H-Heptadecafluoro-1-decanol
Formula:
C₁₀H₅F₁₇O MW: 464.12 g/mol |
MDL Number:
MFCD00039544 CAS Number: 678-39-7 EINECS: 211-648-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...