1H,1H,8H,8H-Dodecafluoro-1,8-octandiol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoro-1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluor-1,8-octandiol, 1H,1H,8H,8H-Perfluoro-1,8-octandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-Dodecafluoro-1,8-octandiol
002305-5G
002305-25G
FLUO002305-5GEA
0
GBP
FLUO002305-5G
FLUO002305-25G
1H,1H,8H,8H-Dodecafluoro-1,8-octandiol
1H,1H,8H,8H-Dodecafluoro-1,8-octandiol
Formula:
C₈H₆F₁₂O₂ MW: 362.12 g/mol |
MDL Number:
MFCD00153145 CAS Number: 90177-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...