1H,1H-Nonadecafluoro-1-decanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluoro-1-decanol, 1H,1H-Perfluoro-1-decanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluor-1-decanol

002297-5G 002297-25G
FLUO002297-5GEA 0 GBP
FLUO002297-5G FLUO002297-25G
1H,1H-Nonadecafluoro-1-decanol
1H,1H-Nonadecafluoro-1-decanol
Formula: C₁₀H₃F₁₉O
MW: 500.1 g/mol
MDL Number: MFCD00153234
CAS Number: 307-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR