1H,1H-Heptadecafluoro-1-nonanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H,1H-Perfluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluor-1-nonanol
002296-5G
002296-25G
FLUO002296-5GEA
0
GBP
FLUO002296-5G
FLUO002296-25G
1H,1H-Heptadecafluoro-1-nonanol
1H,1H-Heptadecafluoro-1-nonanol
Formula:
C₉H₃F₁₇O MW: 450.1 g/mol |
MDL Number:
MFCD00153183 CAS Number: 423-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...