2-Bromo-4-chloro-6-(trifluoromethyl)aniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Bromo-4-chloro-α,α,α-trifluoro-o-toluidine
AN-1276-5G
AN-1276-25G
AN-1276-1G
COBBAN-1276-5GEA
0
GBP
COBBAN-1276-5G
COBBAN-1276-25G
COBBAN-1276-1G
2-Bromo-4-chloro-6-(trifluoromethyl)aniline
2-Bromo-4-chloro-6-(trifluoromethyl)aniline
Formula:
C₇H₄BrClF₃N MW: 274.47 g/mol |
MDL Number:
MFCD08436083 CAS Number: 912617-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...