n-Hexatetracontane, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

52913-250MGEA 63.4 GBP
52913-250MG
n-Hexatetracontane, Supelco®
n-Hexatetracontane
Formula: H₃C(CH₂)₄₄CH₃
MW: 647.25 g/mol
Storage Temperature: Ambient
CAS Number: 7098-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR