2-Propanol

Supplier: VWR Collection
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Isopropyl alcoholIsopropyl rubbing alcohol2-Hydroxy propaneDimethyl carbinolPropan-2-olIPAIsopropanol

VWRS79000GMEA 0 GBP
VWRS79000GM
2-Propanol
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://uk.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg

Order Now

Specification Test Results

Assay Min. 99.5 %
Water Max. 0.1 %
Acidity Max. 0.002 %
Non-volatile residue Max. 0.002 %
Aldehydes + ketones (as CH3COCH3) Max. 0.005 %
Weight per ml 0.784 0.786 g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR