1-Tetradecanol, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Tetradecyl alcohol1-Hydroxy tetradecaneMyristyl alcohol

M-Clarity™ quality level = MQ100

87158-5GEA 190 GBP
87158-5G
1-Tetradecanol, Supelco®
1-Tetradecanol
Formula: H₃C(CH₂)₁₂CH₂OH
MW: 214.39 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 0.8 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004757
CAS Number: 112-72-1
EINECS: 204-000-3
Merck Index: 12,06418

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR