1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1H,1H,2H,2H-Perfluoro-1-iododecane, 1H,1H,2H,2H-Perfluorodecyliodide, 1-Iodo-1H,1H,2H,2H-perfluorodecane
003285-5G
003285-25G
003285-100G
FLUO003285-5GEA
0
GBP
FLUO003285-5G
FLUO003285-25G
FLUO003285-100G
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane
Formula:
C₁₀H₄F₁₇I MW: 574.02 g/mol Boiling Pt: 122 °C (50 mmHg) Melting Pt: 55.5…56.5 °C Density: 1,88 g/cm³ (60 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039411 CAS Number: 2043-53-0 EINECS: 218-053-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...