Caprylic acid

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: n-Octanoic AcidOctoic acidOctanoic acid

EHERCA15711050EA 75.5 GBP
EHERCA15711050
Caprylic acid
Caprylic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR