Propyl acetate

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Acetic acid propyl ester

PROOCHE160EA 287 GBP
PROOCHE160
Propyl acetate
Propyl acetate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0,886 g/cm³ (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00009372
CAS Number: 109-60-4
EINECS: 203-686-1
UN: 1276
ADR: 3,II
Merck Index: 12,08026

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR