trans-4-Aminocyclohexanol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AM-1022-25G AM-1022-100G
COBBAM-1022-25GEA 0 GBP
COBBAM-1022-25G COBBAM-1022-100G
trans-4-Aminocyclohexanol
trans-4-Aminocyclohexanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 127 °C (14 mmHg)
Melting Pt: 108…113 °C
MDL Number: MFCD00067698
CAS Number: 27489-62-9
EINECS: 248-492-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR